(S)-(-)-2-Amino-2-methyl-4-pentenoic acid ≥98%, ee 99%
Lieferant: Thermo Scientific
|
Achtung
|
H54922.MD
H54922.MDEA
149
EUR
H54922.MD
(S)-(-)-2-Amino-2-methyl-4-pentenoic acid ≥98%, ee 99%
(S)-(-)-2-Amino-2-methyl-4-pentenoic acid
Purity: 98%
CAS No.: 96886-55-4
Molecular Formula: C6H11NO2
Formula Weight: 129.16
Storage Temperature: 2°C to 8°C
Melting Point: 285°
Optical Rotation: -14° (c=1.3 in 1N HCl)
MDL No.: MFCD00145248
Formel:
C₆H₁₁NO₂ Molecular Weight: 129,16 g/mol Schmelzpunkt: 285 °C |
MDL:
MFCD00145248 CAS-Nummer: 96886-55-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...