2,4-Difluorobenzylmagnesium bromide 0.25 M in 2-MeTHF (2-Methyltetrahydrofuran)

Lieferant: Thermo Fisher Scientific
Gefahr

H54966.AE
H54966.AEEA 326 EUR
H54966.AE
2,4-Difluorobenzylmagnesium bromide 0.25 M in 2-MeTHF (2-Methyltetrahydrofuran)
2,4-Difluorobenzylmagnesium bromide
Formel: C₇H₅BrF₂Mg
Lagertemperatur: Raumtemperatur
MDL: MFCD07698746
CAS-Nummer: 546122-71-8
UN: 2924
ADR: "3",III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR