2-Amino-9-fluorenon 98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
2-Amino-9H-fluoren-9-one
H55048.03
H55048.06
H55048.14
H55048.03EA
52.1
EUR
H55048.03
H55048.06
H55048.14
2-Amino-9-fluorenon 98%
2-Amino-9-fluorenon
Purity: 98%
CAS No.: 3096-57-9
Molecular Formula: C13H9NO
Formula Weight: 195.22
Melting Point: 157-160°
MDL No.: MFCD00001157
Formel:
C₁₃H₉NO Siedepunkt: 361 °C (1013 hPa) Schmelzpunkt: 157…160 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001157 CAS-Nummer: 3096-57-9 EINECS: 221-445-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...