2,7-Dinitro-9-fluorenon ≥97%
Lieferant: Thermo Fisher Scientific
H55176.06
H55176.14
H55176.06EA
63.9
EUR
H55176.06
H55176.14
2,7-Dinitro-9-fluorenon ≥97%
2,7-Dinitro-9-fluorenon
Formel:
C₁₃H₆N₂O₅ Molecular Weight: 270.2 g/mol Schmelzpunkt: 292…295 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001153 CAS-Nummer: 31551-45-8 EINECS: 250-695-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...