2,7-Dinitro-9-fluorenon ≥97%

Lieferant: Thermo Fisher Scientific

H55176.06 H55176.14
H55176.06EA 63.9 EUR
H55176.06 H55176.14
2,7-Dinitro-9-fluorenon ≥97%
2,7-Dinitro-9-fluorenon
Formel: C₁₃H₆N₂O₅
Molecular Weight: 270.2 g/mol
Schmelzpunkt: 292…295 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00001153
CAS-Nummer: 31551-45-8
EINECS: 250-695-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR