2-Fluor-4-(2-pyridylcarbamoyl)phenylboronsäure ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
2-Fluoro-4-(2-pyridylcarbamoyl)benzeneboronic acid
H55178.03
H55178.03EA
374
EUR
H55178.03
2-Fluor-4-(2-pyridylcarbamoyl)phenylboronsäure ≥97%
2-Fluor-4-(2-pyridylcarbamoyl)phenylboronsäure
Purity: 97%
Molecular Formula: C12H10BFN2O3
Formula Weight: 260.03
MDL No.: MFCD22682831
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...