1-(4-Aminophenyl)piperazine 97%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 4-Piperazin-1-ylaniline

H55224.03
H55224.03EA 71.1 EUR
H55224.03
1-(4-Aminophenyl)piperazine 97%
1-(4-Aminophenyl)piperazine

Purity: 97% CAS No.: 67455-41-8 Molecular Formula: C10H15N3 Formula Weight: 177.25 Physical Form: Powder Appearance: White to purple Melting Point: 119-123° MDL No.: MFCD01365906

Formel: C₁₀H₁₅N₃
Molecular Weight: 177,25 g/mol
Schmelzpunkt: 119…123 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01365906
CAS-Nummer: 67455-41-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR