2-Fluor-6-methylbenzaldehyd ≥97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2-Fluor-6-tolualdehyd, 6-Fluor-o-tolualdehyd, 6-Fluor-o-tolylaldehyd

H55228.03
H55228.03EA 104 EUR
H55228.03
2-Fluor-6-methylbenzaldehyd ≥97%
2-Fluoro-6-methylbenzaldehyde
Formel: C₈H₇FO
Molecular Weight: 138,14 g/mol
Dichte: 1,151
Lagertemperatur: Raumtemperatur
MDL: MFCD09258962
CAS-Nummer: 117752-04-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR