Potassium tetraphenylborate 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H55276.06
H55276.14
H55276.06EA
67.1
EUR
H55276.06
H55276.14
Potassium tetraphenylborate 97%
Potassium tetraphenylborate
Purity: 97%
CAS No.: 3244-41-5
Molecular Formula: C24H20BK
Formula Weight: 358.33
Physical Form: Powder
Appearance: White
Melting Point:
Formel:
C₂₄H₂₀BK Molecular Weight: 358.33 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03093877 CAS-Nummer: 3244-41-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...