2-Chloro-4-(4-morpholinylcarbonyl)benzeneboronic acid 97%
Lieferant: Thermo Scientific
|
Gefahr
|
H55340.03
H55340.03EA
398
EUR
H55340.03
2-Chloro-4-(4-morpholinylcarbonyl)benzeneboronic acid 97%
2-Chloro-4-(4-morpholinylcarbonyl)benzeneboronic acid
Purity: 97%
Molecular Formula: C11H13BClNO4
Formula Weight: 269.49
MDL No.: MFCD22125139
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...