trans,trans-4-(3,4-Difluorophenyl)-4'-n-propylbicyclohexyl 97%

Lieferant: Thermo Fisher Scientific

H55356.03 H55356.06
H55356.03EA 37.1 EUR
H55356.03 H55356.06
trans,trans-4-(3,4-Difluorophenyl)-4'-n-propylbicyclohexyl 97%
trans,trans-4-(3,4-Difluorophenyl)-4'-n-propylbicyclohexyl

Purity: 97% CAS No.: 82832-57-3 Molecular Formula: C21H30F2 Formula Weight: 320.47 Melting Point: 43-46° MDL No.: MFCD09838997

Formel: C₂₁H₃₀F₂
Schmelzpunkt: 43…46 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09838997
CAS-Nummer: 82832-57-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR