Agmatine sulphate 97% (durch HPLC)
Lieferant: Thermo Fisher Scientific
H55363.MD
H55363.03
H55363.06
H55363.MDEA
36.7
EUR
H55363.MD
H55363.03
H55363.06
Agmatine sulphate 97% (durch HPLC)
Agmatine sulphate
Agmatine is an intermediate in the biosynthesis of the polyamines putrescine and spermine.
Agmatine binds with high affinity to α2-adrenergic receptors and imidazoline binding sites and produces blockade of the NMDA-receptor associated cation channels. Agmatine is a competitive inhibitor of nitric oxide synthase activity owing to mimicry of the natural NOS substrate arginine.
Formel:
C₅H₁₄N₄ Schmelzpunkt: 231…238 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00013109 CAS-Nummer: 2482-00-0 EINECS: 219-617-3 Merck Index: 14,188 |
Spezifikation Testergebnisse
Assay (HPLC) | >96.0% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...