Agmatine sulphate 97% (durch HPLC)

Lieferant: Thermo Fisher Scientific

H55363.MD H55363.03 H55363.06
H55363.MDEA 36.7 EUR
H55363.MD H55363.03 H55363.06
Agmatine sulphate 97% (durch HPLC)
Agmatine sulphate

Agmatine is an intermediate in the biosynthesis of the polyamines putrescine and spermine.


Agmatine binds with high affinity to α2-adrenergic receptors and imidazoline binding sites and produces blockade of the NMDA-receptor associated cation channels. Agmatine is a competitive inhibitor of nitric oxide synthase activity owing to mimicry of the natural NOS substrate arginine.

Formel: C₅H₁₄N₄
Schmelzpunkt: 231…238 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00013109
CAS-Nummer: 2482-00-0
EINECS: 219-617-3
Merck Index: 14,188

Bestellen

Spezifikation Testergebnisse

Assay (HPLC) >96.0%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR