4-Cyan-2-iodanilin 97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 4-Amino-3-iodbenzoesäurenitril, 4-Amino-3-iodbenzonitril

H55400.03 H55400.06
H55400.03EA 35.9 EUR
H55400.03 H55400.06
4-Cyan-2-iodanilin 97%
4-Cyan-2-iodanilin

Purity: 97% CAS No.: 33348-34-4 Molecular Formula: C7H5IN2 Formula Weight: 244.03 Melting Point: 112-115° MDL No.: MFCD04039964

Formel: C₇H₅IN₂
Molecular Weight: 244.03 g/mol
Schmelzpunkt: 112…115 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04039964
CAS-Nummer: 33348-34-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR