2,4-Difluorbenzolsulfonamid 96%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2,4-Difluorbenzolsulfonsäureamid
H55437.06
H55437.06EA
302
EUR
H55437.06
2,4-Difluorbenzolsulfonamid 96%
2,4-Difluorbenzolsulfonamid
Purity: 96%
CAS No.: 13656-60-5
Molecular Formula: C6H5F2NO2S
Formula Weight: 193.17
Melting Point: 154-158°
MDL No.: MFCD01320743
Formel:
C₆H₅F₂NO₂S Molecular Weight: 193,17 g/mol Schmelzpunkt: 154…158 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01320743 CAS-Nummer: 13656-60-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...