2,5-Dibromhydrochinon ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 2,5-Dibromobenzene-1,4-diol

H55453.06 H55453.14
H55453.06EA 72.5 EUR
H55453.06 H55453.14
2,5-Dibromhydrochinon ≥97%
2,5-Dibromhydrochinon

Purity: 97% CAS No.: 14753-51-6 Molecular Formula: C6H4Br2O2 Formula Weight: 267.90 Melting Point: 191-194° MDL No.: MFCD00192664

Formel: C₆H₄Br₂O₂
Molecular Weight: 267.9 g/mol
Schmelzpunkt: 191…194 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00192664
CAS-Nummer: 14753-51-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR