(4-Bromophenylethynyl)trimethylsilane 98%
Lieferant: Thermo Fisher Scientific
H55534.03
H55534.06
H55534.14
H55534.03EA
29.2
EUR
H55534.03
H55534.06
H55534.14
(4-Bromophenylethynyl)trimethylsilane 98%
(4-Bromophenylethynyl)trimethylsilane
Purity: 98%
CAS No.: 16116-78-2
Molecular Formula: C11H13BrSi
Formula Weight: 253.21
Physical Form: Powder
Appearance: White to yellow
Melting Point: 61-63°
MDL No.: MFCD01863556
Formel:
C₁₁H₁₃BrSi Schmelzpunkt: 61…63 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01863556 CAS-Nummer: 16116-78-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...