(4-Bromophenylethynyl)trimethylsilane 98%

Lieferant: Thermo Fisher Scientific

H55534.03 H55534.06 H55534.14
H55534.03EA 29.2 EUR
H55534.03 H55534.06 H55534.14
(4-Bromophenylethynyl)trimethylsilane 98%
(4-Bromophenylethynyl)trimethylsilane

Purity: 98% CAS No.: 16116-78-2 Molecular Formula: C11H13BrSi Formula Weight: 253.21 Physical Form: Powder Appearance: White to yellow Melting Point: 61-63° MDL No.: MFCD01863556

Formel: C₁₁H₁₃BrSi
Schmelzpunkt: 61…63 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01863556
CAS-Nummer: 16116-78-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR