2-Chlor-5-formylpyridin 96%

Lieferant: Thermo Fisher Scientific
Achtung

H55550.03 H55550.06
H55550.03EA 94.4 EUR
H55550.03 H55550.06
2-Chlor-5-formylpyridin 96%
2-Chlor-5-formylpyridin

Purity: 96% CAS No.: 23100-12-1 Molecular Formula: C6H4ClNO Formula Weight: 141.56 Melting Point: 77-81° MDL No.: MFCD03095223

Formel: C₆H₄ClNO
Schmelzpunkt: 77…81 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03095223
CAS-Nummer: 23100-12-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR