4-Bromo-N-(4-methoxybenzyl)aniline 97%

Lieferant: Thermo Scientific
Achtung

H55757.03
H55757.03EA 338 EUR
H55757.03
4-Bromo-N-(4-methoxybenzyl)aniline 97%
4-Bromo-N-(4-methoxybenzyl)aniline

Purity: 97% CAS No.: 175357-73-0 Molecular Formula: C14H14NOBr Formula Weight: 292.18 MDL No.: MFCD00017822

Formel: C₁₄H₁₄NOBr
Molecular Weight: 292,18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00017822
CAS-Nummer: 175357-73-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR