2,2'-Dithiodianilin (Thulium-Ionophor I) 97%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Bis(2-aminophenyl)disulfid

H55807.03 H55807.06 H55807.14
H55807.03EA 22 EUR
H55807.03 H55807.06 H55807.14
2,2'-Dithiodianilin (Thulium-Ionophor I) 97%
2,2'-Dithiodianilin (Thulium-Ionophor I)

Purity: 97% CAS No.: 1141-88-4 Molecular Formula: C12H12N2S2 Formula Weight: 248.36 Melting Point: 91-92° Beilstein Reference: 914032

Formel: C₁₂H₁₂N₂S₂
Molecular Weight: 248.37 g/mol
Schmelzpunkt: 91…92 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007703
CAS-Nummer: 1141-88-4
EINECS: 214-529-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR