Di-tert-butyl-L-(+)-tartrate 99%
Lieferant: Thermo Fisher Scientific
Purity: 99%
CAS No.: 117384-45-9
Molecular Formula: C12H22O6
Formula Weight: 262.30
Flash Point: 109°(228°F)
Physical Form: Solid
Density: 1.143
Appearance: White
Melting Point: 90-92°
Beilstein Reference: 5026723
Optical Rotation: +11° (c=1 in acetone)
MDL No.: MFCD00192000
Formel:
C₁₂H₂₂O₆ Schmelzpunkt: 90…92 °C Dichte: 1,143 Flammpunkt: 109 °C (228 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00192000 CAS-Nummer: 117384-45-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...