3,5-Dibromanisol ≥97%

Lieferant: Thermo Scientific

Synonyme: 1,3-Dibromo-5-methoxybenzene

H55925.06
H55925.06EA 175 EUR
H55925.06
3,5-Dibromanisol ≥97%
3,5-Dibromanisol

Purity: 97% CAS No.: 74137-36-3 Molecular Formula: C7H6Br2O Formula Weight: 265.93 Flash Point: 110°(230°F) Melting Point: 34-38° MDL No.: MFCD02258848

Formel: C₇H₆Br₂O
Molecular Weight: 265,93 g/mol
Schmelzpunkt: 34…38 °C
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD02258848
CAS-Nummer: 74137-36-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR