Sodium tetrakis(4-fluorophenyl)borate dihydrate 98% (durch HPLC)

Lieferant: Thermo Fisher Scientific
Achtung

H55952.MD H55952.03 H55952.06
H55952.MDEA 227 EUR
H55952.MD H55952.03 H55952.06
Sodium tetrakis(4-fluorophenyl)borate dihydrate 98% (durch HPLC)
Sodium tetrakis(4-fluorophenyl)borate dihydrate

Sodium tetrakis(4-fluorophenyl)borate dihydrate, is used as a reagent for the two-phase titration of nonionic surfactants in the presence of anionic surfactants.


Bitte beachten Hygroscopic. Incompatible withstrong oxidizing agents.

Formel: C₂₄H₂₀BF₄NaO₂
Molecular Weight: 450.215 g/mol
MDL: MFCD00149598
CAS-Nummer: 207683-22-5

Bestellen

Spezifikation Testergebnisse

Assay (HPLC) Min. 97.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR