Sodium tetrakis(4-fluorophenyl)borate dihydrate 98% (durch HPLC)
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H55952.MD
H55952.03
H55952.06
H55952.MDEA
227
EUR
H55952.MD
H55952.03
H55952.06
Sodium tetrakis(4-fluorophenyl)borate dihydrate 98% (durch HPLC)
Sodium tetrakis(4-fluorophenyl)borate dihydrate
Sodium tetrakis(4-fluorophenyl)borate dihydrate, is used as a reagent for the two-phase titration of nonionic surfactants in the presence of anionic surfactants.
Bitte beachten Hygroscopic. Incompatible withstrong oxidizing agents.
Formel:
C₂₄H₂₀BF₄NaO₂ Molecular Weight: 450.215 g/mol |
MDL:
MFCD00149598 CAS-Nummer: 207683-22-5 |
Spezifikation Testergebnisse
Assay (HPLC) | Min. 97.5% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...