2-Brom-9-fluorenon 96%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 2-Bromo-9H-fluoren-9-one

H55962.MC H55962.ME H55962.14
H55962.MCEA 0 EUR
H55962.MC H55962.ME H55962.14
2-Brom-9-fluorenon 96%
2-Brom-9-fluorenon
Formel: C₁₃H₇BrO
Schmelzpunkt: 146…148 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00010789
CAS-Nummer: 3096-56-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR