4-Chlorphenylboronsäurepinakolester ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
2-(4-Chlorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-Chlorophenylboronic acid Pinacol ester, 4-Chlorobenzeneboronic acid pinacol ester, 1-Chloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene
H55984.03
H55984.06
H55984.14
H55984.03EA
25.7
EUR
H55984.03
H55984.06
H55984.14
4-Chlorphenylboronsäurepinakolester ≥97%
4-Chlorphenylboronsäurepinakolester
Formel:
C₁₂H₁₆BClO₂ Schmelzpunkt: 50…55 °C Flammpunkt: >110 °C (230 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD05663875 CAS-Nummer: 195062-61-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...