4,5-Diazafluoren-9-one 98%

Lieferant: Thermo Fisher Scientific
Achtung

H56028.03 H56028.06
H56028.03EA 132 EUR
H56028.03 H56028.06
4,5-Diazafluoren-9-one 98%
4,5-Diazafluoren-9-one

Purity: 98% CAS No.: 50890-67-0 Molecular Formula: C11H6N2O Formula Weight: 182.18 Melting Point: 214-217° MDL No.: MFCD00046892

Formel: C₁₁H₆N₂O
Schmelzpunkt: 214…217 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00046892
CAS-Nummer: 50890-67-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR