DIPSO 99%

Lieferant: Thermo Fisher Scientific

H56187.22 H56187.30
H56187.22EA 238 EUR
H56187.22 H56187.30
DIPSO 99%
DIPSO

Purity: 99% CAS No.: 68399-80-4 Molecular Formula: C7H17NO6S Formula Weight: 261.29 Beilstein Reference: 2105454 MDL No.: MFCD00038353

Formel: C₇H₁₇NO₆S
Lagertemperatur: Raumtemperatur
MDL: MFCD00038353
CAS-Nummer: 68399-80-4
EINECS: 269-992-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR