2-Brom-3-hexylthiophen 98%

Lieferant: Thermo Fisher Scientific
Achtung

H56243.06
H56243.06EA 131 EUR
H56243.06
2-Brom-3-hexylthiophen 98%
2-Bromo-3-hexylthiophene

Purity: 98% CAS No.: 69249-61-2 Molecular Formula: C10H15BrS Formula Weight: 247.20 Flash Point: 110°(230°F) Refractive Index: 1.529 MDL No.: MFCD09907959

Formel: C₁₀H₁₅BrS
Dichte: 1,240
Flammpunkt: 110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD09907959
CAS-Nummer: 69249-61-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR