5-Brom-4-chlor-3-indolylphosphat Dinatriumsalz (BCIP-Na) ≥98%
Lieferant: Thermo Fisher Scientific
Synonyme:
5-Bromo-4-chloro-1H-indol-3-yl phosphate sodium, X-phosphate disodium salt, Sodium 5-bromo-4-chloro-1H-indol-3-yl phosphate
H56265.03
H56265.03EA
355
EUR
H56265.03
5-Brom-4-chlor-3-indolylphosphat Dinatriumsalz (BCIP-Na) ≥98%
5-Brom-4-chlor-3-indolylphosphat Dinatriumsalz (BCIP-Na)
Purity: 98+%
CAS No.: 102185-33-1
Molecular Formula: C8H4BrClNNa2O4P
Formula Weight: 370.43
Storage Temperature: -30°C to -10°C
Formel:
C₈H₄BrClNNa₂O₄P |
MDL:
MFCD00036757 CAS-Nummer: 102185-33-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...