4-Iod-1H-pyrazol ≥99%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 4-Iodopyrazole

H56369.06 H56369.14
H56369.06EA 104 EUR
H56369.06 H56369.14
4-Iod-1H-pyrazol ≥99%
4-Iodo-1H-pyrazole
Formel: C₃H₃IN₂
Molecular Weight: 193.97 g/mol
Schmelzpunkt: 108…110 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00005244
CAS-Nummer: 3469-69-0
EINECS: 222-434-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR