4-Iod-1H-pyrazol ≥99%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
4-Iodopyrazole
H56369.06
H56369.14
H56369.06EA
104
EUR
H56369.06
H56369.14
4-Iod-1H-pyrazol ≥99%
4-Iodo-1H-pyrazole
Formel:
C₃H₃IN₂ Molecular Weight: 193.97 g/mol Schmelzpunkt: 108…110 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005244 CAS-Nummer: 3469-69-0 EINECS: 222-434-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...