β-(N-Morpholino)ethanesulphonic acid (MES) hydrate ≥99%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
4-Morpholineethanesulphonic acid hydrate, MES hydrate, 2-(4-Morpholinyl)ethanesulphonic acid hydrate
H56472.36
H56472.36EA
321
EUR
H56472.36
β-(N-Morpholino)ethanesulphonic acid (MES) hydrate ≥99%
β-(N-Morpholino)ethanesulphonic acid (MES) hydrate
MES hydrate is a zwitterionic N-substituted aminosulfonic acid, known as Good's buffer, is utilized for many biological applications. It is a useful resolving agent for very small proteins on bis-tris gels. It is mainly used as a biological buffer in plant cell cultures. Buffer solutions are used as a means of keeping pH at a nearly constant value in a wide variety of chemical applications.
Formel:
C₆H₁₃NO₄S·xH₂O Molecular Weight: 195,24 g/mol (anhy) Schmelzpunkt: ∼ 308 °C (Zersetzung) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149409 CAS-Nummer: 1266615-59-1 EINECS: 224-632-3 Merck Index: 14,5902 |
Spezifikation Testergebnisse
Appearance (colour) | White |
Assay | 99.0% |
Water Content (Karl Fischer Titration): |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...