4-Carboxy-1-naphthalinboronsäure 95%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 4-Borono-1-naphthoic acid

H56542.MD H56542.03
H56542.MDEA 176 EUR
H56542.MD H56542.03
4-Carboxy-1-naphthalinboronsäure 95%
4-Carboxy-1-naphthalinboronsäure
Formel: C₁₁H₉BO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD06203538
CAS-Nummer: 332398-57-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR