5-Methylphenaziniummethylsulfat ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H56718.06
H56718.14
H56718.06EA
175
EUR
H56718.06
H56718.14
5-Methylphenaziniummethylsulfat ≥98%
5-Methylphenazinium methyl sulphate
Purity: 98+%
CAS No.: 299-11-6
Molecular Formula: C13H11N2.CH3SO4
Formula Weight: 306.34
Storage Temperature: -30°C to -10°C
Formel:
C₁₄H₁₄N₂O₄S Schmelzpunkt: 198 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD00011923 CAS-Nummer: 299-11-6 EINECS: 206-072-1 Merck Index: 12,06185 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...