Methyl-4-hydroxy-3-iodobenzoate 98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Methyl 4-hydroxy-3-iodobenzoate
H56877.06
H56877.06EA
77.6
EUR
H56877.06
Methyl-4-hydroxy-3-iodobenzoate 98%
Methyl-4-hydroxy-3-iodobenzoate
Purity: 98%
CAS No.: 15126-06-4
Molecular Formula: C8H7IO3
Formula Weight: 278.04
Physical Form: Crystal or powder
Sensitivity: Light Sensitive
Appearance: White to pale brown
Melting Point: 155-159°
MDL No.: MFCD05664378
Formel:
C₈H₇IO₃ Molecular Weight: 278,04 g/mol Schmelzpunkt: 155…159 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD05664378 CAS-Nummer: 15126-06-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...