Ethyl (R)-(+)-1-boc-4-oxopiperidine-2-carboxylate 95%

Lieferant: Thermo Fisher Scientific
Achtung

H57019.03
H57019.03EA 235 EUR
H57019.03
Ethyl (R)-(+)-1-boc-4-oxopiperidine-2-carboxylate 95%
Ethyl (R)-(+)-1-boc-4-oxopiperidine-2-carboxylate

Purity: 95% CAS No.: 357154-16-6 Molecular Formula: C13H21NO5 Formula Weight: 271.31 Storage Temperature: 2°C to 8°C Optical Rotation: +20° (c=0.5 in chloroform) MDL No.: MFCD20526405

Formel: C₁₃H₂₁NO₅
MDL: MFCD20526405
CAS-Nummer: 357154-16-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR