6-Boc-1-oxa-6-azaspiro[3.3]heptane 96%

Lieferant: Thermo Scientific
Achtung

H57040.MD
H57040.MDEA 112 EUR
H57040.MD
6-Boc-1-oxa-6-azaspiro[3.3]heptane 96%
6-Boc-1-oxa-6-azaspiro[3.3]heptane

Purity: 96% CAS No.: 1223573-41-8 Molecular Formula: C10H17NO3 Formula Weight: 199.25

Formel: C₁₀H₁₇NO₃
Molecular Weight: 199,25 g/mol
Lagertemperatur: Raumtemperatur
CAS-Nummer: 1223573-41-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR