tert-Butyl-3-bromopyrrolidine-1-carboxylate 95%

Lieferant: Thermo Scientific
Achtung

Synonyme: (±)-1-Boc-3-bromopyrrolidine

H57071.06
H57071.06EA 614 EUR
H57071.06
tert-Butyl-3-bromopyrrolidine-1-carboxylate 95%
tert-Butyl-3-bromopyrrolidine-1-carboxylate

Purity: 95% CAS No.: 939793-16-5 Molecular Formula: C9H16BrNO2 Formula Weight: 250.14 Storage Temperature: 2°C to 8°C Melting Point: 45-53° UN No.: UN3077 MDL No.: MFCD08752478

Formel: C₉H₁₆BrNO₂
Molecular Weight: 250,14 g/mol
Schmelzpunkt: 45…53 °C
MDL: MFCD08752478
CAS-Nummer: 939793-16-5
ADR: 9,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR