7-Phenyl-1,4-diazepan-5-one 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 7-Phenylhomopiperazin-5-one

H57126.03
H57126.03EA 122 EUR
H57126.03
7-Phenyl-1,4-diazepan-5-one 97%
7-Phenyl-1,4-diazepan-5-one

Purity: 97% CAS No.: 89044-79-1 Molecular Formula: C11H14N2O Formula Weight: 190.24 Melting Point: 104-106° MDL No.: MFCD12197004

Formel: C₁₁H₁₄N₂O
Molecular Weight: 190,25 g/mol
Schmelzpunkt: 104…106 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD12197004
CAS-Nummer: 89044-79-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR