2-Chloro-N,N-dimethylnicotinamide 97%

Lieferant: Thermo Fisher Scientific
Achtung

H57162.MD H57162.03
H57162.MDEA 52.8 EUR
H57162.MD H57162.03
2-Chloro-N,N-dimethylnicotinamide 97%
2-Chloro-N,N-dimethylnicotinamide

Purity: 97% CAS No.: 52943-21-2 Molecular Formula: C8H9ClN2O Formula Weight: 184.63 Melting Point: 63-65° MDL No.: MFCD08593346

Formel: C₈H₉ClN₂O
Schmelzpunkt: 63…65 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08593346
CAS-Nummer: 52943-21-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR