Pyrrolidin-3-ylmethanol ≥97%
Lieferant: Thermo Scientific
|
Achtung
|
H57225.03
H57225.03EA
274
EUR
H57225.03
Pyrrolidin-3-ylmethanol ≥97%
Pyrrolidin-3-ylmethanol
Purity: 97+%
CAS No.: 5082-74-6
Molecular Formula: C5H11NO
Formula Weight: 101.15
Storage Temperature: 2°C to 8°C
Boiling Point: 165-171°
Beilstein Reference: 1420904
MDL No.: MFCD07773080
Formel:
C5H11NO Molecular Weight: 101,15 g/mol Siedepunkt: 165…171 °C Dichte: 0,979 |
MDL:
MFCD07773080 CAS-Nummer: 5082-74-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...