2-Methyl-N-(3-oxetanylidene)propane-2-sulphinamide 95%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H57776.ME
H57776.MEEA
228
EUR
H57776.ME
2-Methyl-N-(3-oxetanylidene)propane-2-sulphinamide 95%
2-Methyl-N-(3-oxetanylidene)propane-2-sulphinamide
Purity: 95%
CAS No.: 1158098-73-7
Molecular Formula: C7H13NO2S
Formula Weight: 175.25
MDL No.: MFCD14702522
Formel:
C₇H₁₃NO₂S Lagertemperatur: Raumtemperatur |
MDL:
MFCD14702522 CAS-Nummer: 1158098-73-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...