L-β-Proline ethyl ester hydrochloride 97%
Lieferant: Thermo Scientific
|
Achtung
|
H57781.03
H57781.03EA
234
EUR
H57781.03
L-β-Proline ethyl ester hydrochloride 97%
L-beta-Proline ethyl ester hydrochloride
Purity: 97%
Molecular Formula: C7H13NO2.HCl
Formula Weight: 179.65
Storage Temperature: 2°C to 8°C
Optical Rotation: +14° (c=0.6 in water)
MDL No.: MFCD21337911
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...