D-β-Proline ethyl ester hydrochloride 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H57789.03
H57789.03EA
126
EUR
H57789.03
D-β-Proline ethyl ester hydrochloride 97%
D-beta-Proline ethyl ester hydrochloride
Purity: 97%
Molecular Formula: C7H13NO2.HCl
Formula Weight: 179.65
Storage Temperature: 2°C to 8°C
Optical Rotation: -14° (c=0.6 in water)
MDL No.: MFCD18071037