(R)-1-(tert-Butoxycarbonyl)pyrrolidine-3-carboxylic acid 95%
Lieferant: Thermo Scientific
|
Achtung
|
H57825.03
H57825.03EA
156
EUR
H57825.03
(R)-1-(tert-Butoxycarbonyl)pyrrolidine-3-carboxylic acid 95%
(R)-1-(tert-Butoxycarbonyl)pyrrolidine-3-carboxylic acid
Purity: 95%
CAS No.: 72925-16-7
Molecular Formula: C10H17NO4
Formula Weight: 215.25
Storage Temperature: 2°C to 8°C
Optical Rotation: -15° (c=0.8 in chloroform)
MDL No.: MFCD03094727
Formel:
C₁₀H₁₇NO₄ Molecular Weight: 215,25 g/mol |
MDL:
MFCD03094727 CAS-Nummer: 72925-16-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...