2-Oxa-5-azaspiro[3.4]octane oxalate 96%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H57854.MD
H57854.ME
H57854.MDEA
399
EUR
H57854.MD
H57854.ME
2-Oxa-5-azaspiro[3.4]octane oxalate 96%
2-Oxa-5-azaspiro[3.4]octane oxalate
Purity: 96%
CAS No.: 1389264-18-9
Molecular Formula: C6H11NO.C2H2O4
Formula Weight: 203.19
Storage Temperature: 2°C to 8°C
MDL No.: MFCD18325162
Formel:
C₆H₁₁NO·C₂H₂O₄ Molecular Weight: 203.19 g/mol |
MDL:
MFCD18325162 CAS-Nummer: 1389264-18-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...