Ethyl-2-oxopyrrolidine-3-carboxylate 95%

Lieferant: Thermo Scientific
Achtung

Synonyme: Ethyl 2-oxo-pyrrolidine-3-carboxylate

H57906.03
H57906.03EA 163 EUR
H57906.03
Ethyl-2-oxopyrrolidine-3-carboxylate 95%
Ethyl-2-oxopyrrolidine-3-carboxylate

Purity: 95% CAS No.: 36821-26-8 Molecular Formula: C7H11NO3 Formula Weight: 157.17 Storage Temperature: 2°C to 8°C Melting Point: 71-75° MDL No.: MFCD06796534

Formel: C₇H₁₁NO₃
Molecular Weight: 157,17 g/mol
Schmelzpunkt: 71…75 °C
MDL: MFCD06796534
CAS-Nummer: 36821-26-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR