3-Oxetanol 95%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
H57928.06
H57928.03
H57928.06EA
125
EUR
H57928.06
H57928.03
3-Oxetanol 95%
Oxetan-3-ol
Purity: 95%
CAS No.: 7748-36-9
Molecular Formula: C3H6O2
Formula Weight: 74.08
Flash Point: 87°(189°F)
Refractive Index: 1.438
Boiling Point: 153°
MDL No.: MFCD09056790
Formel:
C₃H₆O₂ Siedepunkt: 153 °C Dichte: 1,167 Flammpunkt: 87 °C (189 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD09056790 CAS-Nummer: 7748-36-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...